Assay Details
Gene Symbol: Cdca7l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034953.1 | NP_001030125.1 | 1 - 2 | 76 | 99 | |
GenBank mRNA | BC098734.1 | - | 1 - 2 | 76 | 7385105 | 99 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC098734.1,NM_001034953.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
