Assay Details
Gene Symbol: Cdca7l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034953.1 | NP_001030125.1 | 2 - 3 | 204 | 138 | |
GenBank mRNA | BC098734.1 | - | 2 - 3 | 204 | 138 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
