Assay Details
Gene Symbol: Cdca7l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034953.1 | NP_001030125.1 | 5 - 6 | 765 | 114 | |
GenBank mRNA | BC098734.1 | - | 5 - 6 | 765 | 7385105 | 114 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
