Assay Details
Gene Symbol: Cdca7l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034953.1 | NP_001030125.1 | 8 - 9 | 1211 | 108 | |
GenBank mRNA | BC098734.1 | - | 8 - 9 | 1211 | 7385105 | 108 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P038887 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600697 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 98497 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473893 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850618 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289972 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373222 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834841 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105560 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213145 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184885 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189055 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808442 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858650 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232889 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872531 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 123555 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599994 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 113902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404546 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445285 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10108 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50660 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357394 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782928 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908440 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46691 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928088 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795901 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32573 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21194 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763127 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70317 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4097 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 34782 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12245 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159292 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P038887 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P038887 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P038887 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 55122 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 55556 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 11869 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 9452 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 103226 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 59653 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10840652 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887805 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887806 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10840647 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887807 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887804 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887803 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887808 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10840640 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10887802 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10840650 | - | - |
Rat Genome 230 2.0 Array | 1389755_at | - | - |
Rat Genome 230 2.0 Array | 1389755_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1389755_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1389755_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0600012095.rn.1 | TC0600001701.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600012096.rn.1 | TC0600001701.rn.1 | RTA TC |
Rat Expression Set 230 | 1389755_PM_at | - | - |
Rat Expression Set 230 | 1389755_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887802 | - | - |
Rat Genome U34 Set | rc_AI071549_at | - | - |
Rat Genome U34 Set | rc_AI178707_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21447495 | - | - |
