Assay Details
Gene Symbol: Cirbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006241012.2 | XP_006241074.1 | 4 - 5 | 525 | 141 | |
XM_006241013.2 | XP_006241075.2 | 4 - 5 | 636 | 141 | ||
XM_006241014.2 | XP_006241076.1 | 4 - 5 | 525 | 141 | ||
XM_006241015.2 | XP_006241077.2 | 4 - 5 | 636 | 141 | ||
XM_008765132.1 | XP_008763354.1 | 4 - 5 | 636 | 141 | ||
GenBank mRNA | AB000362.1 | - | 3 - 4 | 369 | 141 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
