Assay Details
Gene Symbol: Cirbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008765132.1 | XP_008763354.1 | 6 - 7 | 789 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
