Assay Details
Gene Symbol: Apba3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031781.1 | NP_113969.1 | 7 - 8 | 1465 | 93 | |
XM_006241018.2 | XP_006241080.1 | 8 - 9 | 1626 | 93 | ||
XM_006241019.2 | XP_006241081.1 | 8 - 9 | 1599 | 93 | ||
XM_006241020.2 | XP_006241082.1 | 8 - 9 | 1624 | 93 | ||
GenBank mRNA | AF029109.1 | - | 7 - 8 | 1465 | 93 | |
BC161842.1 | - | 8 - 9 | 1527 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
