Assay Details
Gene Symbol: Stk11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108069.1 | NP_001101539.1 | 2 - 3 | 1201 | 66 | |
XM_006240910.1 | XP_006240972.1 | 2 - 3 | 1196 | 66 | ||
XM_008765096.1 | XP_008763318.1 | 2 - 3 | 1196 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
