Assay Details
Gene Symbol: Shc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108065.1 | NP_001101535.1 | 12 - 13 | 1798 | 151 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P124567 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290582 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 666920 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569751 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907912 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911319 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441057 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9261 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518149 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223251 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960080 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119171 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 68897 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407369 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312697 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212942 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652816 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 68709 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27212 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82967 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P124567 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P124567 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P124567 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P485521 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 73660 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10894061 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10894069 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10894056 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10894062 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10894068 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10894059 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10894063 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10894058 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10894060 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10894057 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10894064 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10894066 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10894065 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10894055 | - | - |
Rat Genome 230 2.0 Array | 1384347_at | - | - |
Rat Genome 230 2.0 Array | 1378693_PM_at | - | - |
Rat Genome 230 2.0 Array | 1378693_at | - | - |
Rat Genome 230 2.0 Array | 1384347_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1378693_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1378693_at | - | - |
Affymetrix HT Rat Focus Array | 1384347_at | - | - |
Affymetrix HT Rat Focus Array | 1384347_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0700002272.rn.1 | TC0700000237.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700002271.rn.1 | TC0700000237.rn.1 | RTA TC |
Rat Expression Set 230 | 1384347_PM_at | - | - |
Rat Expression Set 230 | 1384347_at | - | - |
Rat Expression Set 230 | 1378693_at | - | - |
Rat Expression Set 230 | 1378693_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10894055 | - | - |
Rat Genome U34 Set | AB001452_at | - | - |
Rat Genome U34 Set | rc_AA859760_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21816631 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1357475 | - | - |
