Assay Details
Gene Symbol: Bcl2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033672.1 | NP_001028844.1 | 1 - 2 | 227 | 75 | |
XM_006235265.2 | XP_006235327.1 | 1 - 2 | 882 | 75 | ||
GenBank mRNA | AY141038.1 | - | 1 - 2 | 227 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AY141038.1, NM_001033672.1, XM_006235265.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
