Assay Details
Gene Symbol: Uqcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109446.2 | NP_001102916.1 | 8 - 9 | 652 | 90 | |
XM_006235361.2 | XP_006235423.1 | 8 - 9 | 726 | 90 | ||
GenBank mRNA | BC107807.1 | - | 8 - 9 | 652 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
