Assay Details
Gene Symbol: Pigu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181637.1 | NP_853668.1 | 10 - 11 | 1057 | 82 | |
XM_006235325.2 | XP_006235387.1 | 11 - 12 | 1308 | 82 | ||
XM_008762347.1 | XP_008760569.1 | 8 - 9 | 1405 | 82 | ||
GenBank mRNA | AB086841.1 | - | 10 - 11 | 1092 | 82 | |
BC097269.1 | - | 11 - 12 | 1183 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P043456 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869555 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382496 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273520 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 219122 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 584735 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491851 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825637 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 645753 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589952 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726248 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860825 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775025 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513689 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79596 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426631 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413312 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184720 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510328 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 4211 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 71103 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831401 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902244 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948016 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652844 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763148 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61387 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669631 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 551532 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379879 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796857 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154104 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812238 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741661 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571080 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86476 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524795 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745084 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550641 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 199334 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942353 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413633 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535530 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 219057 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939227 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305528 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134165 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878002 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808114 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133952 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121075 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244064 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451101 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169086 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 110180 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 3249 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106249 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 172587 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55057 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 25483 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79308 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39853 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140579 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P043456 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P043456 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P043456 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P515257 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 57546 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 41501 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 23140 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 13302 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 88076 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 89012 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 9618 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10850996 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10850991 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10850992 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10850997 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10850998 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10850994 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10851002 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10850995 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10850993 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10850990 | - | - |
Rat Genome 230 2.0 Array | 1374357_PM_at | - | - |
Rat Genome 230 2.0 Array | 1374357_at | - | - |
Affymetrix HT Rat Focus Array | 1374357_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1374357_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0300039488.rn.1 | TC0300005100.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300039490.rn.1 | TC0300005100.rn.1 | RTA TC |
Rat Expression Set 230 | 1374357_at | - | - |
Rat Expression Set 230 | 1374357_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10850990 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21727996 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1370471 | - | - |
RatRef-12 Expression BeadChip | ILMN_63884 | - | - |
