Assay Details
Gene Symbol: Pigu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181637.1 | NP_853668.1 | 6 - 7 | 530 | 104 | |
XM_006235325.2 | XP_006235387.1 | 7 - 8 | 781 | 104 | ||
XM_008762347.1 | XP_008760569.1 | 4 - 5 | 878 | 104 | ||
GenBank mRNA | AB086841.1 | - | 6 - 7 | 565 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
