Assay Details
Gene Symbol: Rbm39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013207.2 | NP_001013225.1 | 358 | 116 | ||
XM_006235331.2 | XP_006235393.1 | 416 | 116 | |||
XM_006235332.2 | XP_006235394.1 | 437 | 116 | |||
XM_008762349.1 | XP_008760571.1 | 419 | 116 | |||
XM_008762350.1 | XP_008760572.1 | 440 | 116 | |||
XM_008762356.1 | XP_008760578.1 | 419 | 116 | |||
GenBank mRNA | BC078917.1 | - | 431 | 116 | ||
BC091394.1 | - | 347 | 116 |
Gene Symbol: LOC100910882 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749605.3 | XP_003749653.1 | 3 - 4 | 416 | 116 | |
XM_003749606.3 | XP_003749654.1 | 3 - 4 | 416 | 116 | ||
XM_006235443.2 | XP_006235505.1 | 3 - 4 | 437 | 116 | ||
XM_008762415.1 | XP_008760637.1 | 3 - 4 | 419 | 116 | ||
XM_008762416.1 | XP_008760638.1 | 3 - 4 | 440 | 116 | ||
XM_008762422.1 | XP_008760644.1 | 3 - 4 | 419 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
