Assay Details
Gene Symbol: Rbm39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013207.2 | NP_001013225.1 | 252 | 117 | ||
XM_006235331.2 | XP_006235393.1 | 310 | 117 | |||
XM_008762349.1 | XP_008760571.1 | 313 | 117 | |||
XM_008762355.1 | XP_008760577.1 | 309 | 190 | |||
XM_008762356.1 | XP_008760578.1 | 313 | 117 | |||
GenBank mRNA | BC078917.1 | - | 252 | 190 | ||
BC091394.1 | - | 241 | 117 |
Gene Symbol: LOC100910882 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749605.3 | XP_003749653.1 | 1 - 2 | 310 | 117 | |
XM_003749606.3 | XP_003749654.1 | 1 - 2 | 310 | 117 | ||
XM_008762415.1 | XP_008760637.1 | 1 - 2 | 313 | 117 | ||
XM_008762421.1 | XP_008760643.1 | 1 - 2 | 309 | 190 | ||
XM_008762422.1 | XP_008760644.1 | 1 - 2 | 313 | 117 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC091394.1, NM_001013207.2, XM_003749605.3, XM_003749606.3, XM_006235331.2, XM_008762349.1, XM_008762355.1, XM_008762356.1, XM_008762415.1, XM_008762421.1, XM_008762422.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
