Assay Details
Gene Symbol: Rbm39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013207.2 | NP_001013225.1 | 1551 | 159 | ||
XM_006235331.2 | XP_006235393.1 | 1627 | 159 | |||
XM_006235332.2 | XP_006235394.1 | 1648 | 159 | |||
XM_006235335.2 | XP_006235397.1 | 3069 | 159 | |||
XM_008762349.1 | XP_008760571.1 | 1564 | 159 | |||
XM_008762350.1 | XP_008760572.1 | 1585 | 159 | |||
XM_008762351.1 | XP_008760573.1 | 13 - 14 | 7467 | 159 | ||
XM_008762352.1 | XP_008760574.1 | 13 - 14 | 4559 | 159 | ||
XM_008762353.1 | XP_008760575.1 | 6201 | 159 | |||
XM_008762354.1 | XP_008760576.1 | 13 - 14 | 7488 | 159 | ||
XM_008762355.1 | XP_008760577.1 | 5977 | 159 | |||
XM_008762356.1 | XP_008760578.1 | 1546 | 159 | |||
GenBank mRNA | AY769432.1 | - | 421 | 159 | ||
BC078917.1 | - | 1642 | 159 | |||
BC091394.1 | - | 1540 | 159 |
Gene Symbol: LOC100910882 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749605.3 | XP_003749653.1 | 14 - 15 | 1627 | 159 | |
XM_003749606.3 | XP_003749654.1 | 14 - 15 | 1609 | 159 | ||
XM_006235443.2 | XP_006235505.1 | 14 - 15 | 1648 | 159 | ||
XM_006235446.2 | XP_006235508.1 | 14 - 15 | 3069 | 159 | ||
XM_008762415.1 | XP_008760637.1 | 13 - 14 | 1564 | 159 | ||
XM_008762416.1 | XP_008760638.1 | 13 - 14 | 1585 | 159 | ||
XM_008762417.1 | XP_008760639.1 | 13 - 14 | 7467 | 159 | ||
XM_008762418.1 | XP_008760640.1 | 13 - 14 | 4559 | 159 | ||
XM_008762419.1 | XP_008760641.1 | 13 - 14 | 6201 | 159 | ||
XM_008762420.1 | XP_008760642.1 | 13 - 14 | 7488 | 159 | ||
XM_008762421.1 | XP_008760643.1 | 14 - 15 | 5977 | 159 | ||
XM_008762422.1 | XP_008760644.1 | 13 - 14 | 1546 | 159 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
