Assay Details
Gene Symbol: Raly | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011958.1 | NP_001011958.1 | 1 - 2 | 67 | 88 | |
XM_008762314.1 | XP_008760536.1 | 1 - 2 | 457 | 88 | ||
GenBank mRNA | BC087074.1 | - | 1 - 2 | 67 | 7134486 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC087074.1, NM_001011958.1, XM_008762314.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
