Assay Details
Gene Symbol: Raly | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008762314.1 | XP_008760536.1 | 4 - 5 | 878 | 78 | |
XM_008762315.1 | XP_008760537.1 | 3 - 4 | 596 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
