Assay Details
Gene Symbol: Cbfa2t2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168542.1 | NP_001162014.1 | 1 - 2 | 154 | 89 | |
XM_006235311.2 | XP_006235373.1 | 1 - 2 | 138 | 89 | ||
GenBank mRNA | FQ222452.1 | - | 1 - 2 | 170 | 89 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001168542.1, XM_006235311.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
