Assay Details
Gene Symbol: Tbc1d20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004281.1 | NP_001004281.1 | 1 - 2 | 118 | 104 | |
XM_008762348.1 | XP_008760570.1 | 1 - 2 | 267 | 104 | ||
GenBank mRNA | BC079383.1 | - | 1 - 2 | 118 | 7111239 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
