Assay Details
Gene Symbol: Tbc1d20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004281.1 | NP_001004281.1 | 5 - 6 | 681 | 112 | |
XM_008762348.1 | XP_008760570.1 | 5 - 6 | 830 | 112 | ||
GenBank mRNA | BC079383.1 | - | 5 - 6 | 681 | 7111239 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
