Assay Details
Gene Symbol: Tbc1d20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004281.1 | NP_001004281.1 | 6 - 7 | 816 | 60 | |
GenBank mRNA | BC079383.1 | - | 6 - 7 | 816 | 7111239 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
