Assay Details
Gene Symbol: Sla2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172118.1 | NP_001165589.1 | 4 - 5 | 387 | 125 | |
XM_008762374.1 | XP_008760596.1 | 6 - 7 | 2064 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
