Assay Details
Gene Symbol: Dusp15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244784.1 | NP_001231713.1 | 5 - 6 | 465 | 91 | |
GenBank mRNA | BC168211.1 | - | 5 - 6 | 294 | 7135378 | 91 |
CB585518.1 | - | 5 - 6 | 465 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
