Assay Details
Gene Symbol: Dusp15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108598.2 | NP_001102068.2 | 2 - 3 | 262 | 85 | |
NM_001244784.1 | NP_001231713.1 | 2 - 3 | 262 | 85 | ||
GenBank mRNA | BC168211.1 | - | 2 - 3 | 91 | 85 | |
CB576376.1 | - | 2 - 3 | 139 | 85 | ||
CB585518.1 | - | 2 - 3 | 262 | 85 | ||
CB736780.1 | - | 2 - 3 | 341 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
