Assay Details
Gene Symbol: Dynlrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131910.3 | NP_571985.1 | 1 - 2 | 127 | 60 | |
GenBank mRNA | AF073839.2 | - | 1 - 2 | 50 | 60 | |
AY026512.1 | - | 1 - 2 | 127 | 60 | ||
BC058437.1 | - | 1 - 2 | 68 | 6921076 | 60 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF073839.2, AY026512.1, BC058437.1, NM_131910.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
