Assay Details
Gene Symbol: Cdh8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053393.2 | NP_445845.2 | 10 - 11 | 2188 | 177 | |
XM_008772244.1 | XP_008770466.1 | 9 - 10 | 1535 | 177 | ||
GenBank mRNA | AB010436.1 | - | 10 - 11 | 2188 | 177 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
