Assay Details
Gene Symbol: Cdh8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053393.2 | NP_445845.2 | 1 - 2 | 334 | 92 | |
GenBank mRNA | AB010436.1 | - | 1 - 2 | 334 | 92 | |
AB010437.1 | - | 1 - 2 | 334 | 92 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AB010436.1, AB010437.1, NM_053393.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
