Assay Details
Gene Symbol: Cdh8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053393.2 | NP_445845.2 | 4 - 5 | 1193 | 72 | |
XM_008772244.1 | XP_008770466.1 | 3 - 4 | 540 | 72 | ||
GenBank mRNA | AB010436.1 | - | 4 - 5 | 1193 | 72 | |
AB010437.1 | - | 4 - 5 | 1193 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
