Assay Details
Gene Symbol: Cdh8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053393.2 | NP_445845.2 | 7 - 8 | 1808 | 82 | |
XM_008772244.1 | XP_008770466.1 | 6 - 7 | 1155 | 82 | ||
GenBank mRNA | AB010436.1 | - | 7 - 8 | 1808 | 82 | |
AB010437.1 | - | 7 - 8 | 1808 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
