Assay Details
Gene Symbol: Dync1li2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031026.1 | NP_112288.1 | 10 - 11 | 1222 | 76 | |
XM_006255053.2 | XP_006255115.1 | 11 - 12 | 1478 | 76 | ||
XM_006255054.2 | XP_006255116.1 | 10 - 11 | 1436 | 76 | ||
GenBank mRNA | AB008521.1 | - | 6 - 7 | 530 | 76 | |
AB008522.1 | - | 5 - 6 | 488 | 76 | ||
AB008523.1 | - | 5 - 6 | 488 | 76 | ||
AB008524.1 | - | 6 - 7 | 530 | 76 | ||
BC089794.1 | - | 11 - 12 | 1285 | 76 | ||
U15138.1 | - | 10 - 11 | 1222 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
