Assay Details
Gene Symbol: Dync1li2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031026.1 | NP_112288.1 | 6 - 7 | 801 | 168 | |
XM_006255053.2 | XP_006255115.1 | 6 - 7 | 1015 | 168 | ||
XM_006255054.2 | XP_006255116.1 | 6 - 7 | 1015 | 168 | ||
GenBank mRNA | AB008521.1 | - | 1 - 2 | 67 | 168 | |
AB008522.1 | - | 1 - 2 | 67 | 168 | ||
AB008523.1 | - | 1 - 2 | 67 | 168 | ||
AB008524.1 | - | 1 - 2 | 67 | 168 | ||
BC089794.1 | - | 6 - 7 | 822 | 168 | ||
U15138.1 | - | 6 - 7 | 801 | 168 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
