Assay Details
Gene Symbol: Nudt21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039004.1 | NP_001034093.1 | 4 - 5 | 557 | 99 | |
XM_006255132.2 | XP_006255194.1 | 4 - 5 | 1190 | 99 | ||
GenBank mRNA | BC098748.1 | - | 4 - 5 | 557 | 99 | |
BC168149.1 | - | 4 - 5 | 1196 | 99 | ||
FQ229498.1 | - | 4 - 5 | 598 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
