Assay Details
Gene Symbol: Herpud1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053523.1 | NP_445975.1 | 5 - 6 | 647 | 72 | |
XM_006255133.2 | XP_006255195.1 | 5 - 6 | 960 | 72 | ||
GenBank mRNA | AB033771.1 | - | 5 - 6 | 647 | 72 | |
BC061849.1 | - | 5 - 6 | 669 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
