Assay Details
Gene Symbol: Herpud1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053523.1 | NP_445975.1 | 6 - 7 | 996 | 139 | |
XM_006255133.2 | XP_006255195.1 | 6 - 7 | 1309 | 139 | ||
GenBank mRNA | AB033771.1 | - | 6 - 7 | 996 | 139 | |
BC061849.1 | - | 6 - 7 | 1018 | 139 | ||
FQ219047.1 | - | 1 - 2 | 141 | 139 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
