Assay Details
Gene Symbol: Cdh5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107407.1 | NP_001100877.1 | 8 - 9 | 1396 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
