Assay Details
Gene Symbol: Setd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106167.1 | NP_001099637.1 | 1 - 2 | 362 | 70 | |
XM_006255070.2 | XP_006255132.1 | 1 - 2 | 1240 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
