Assay Details
Gene Symbol: Polr2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012473.1 | NP_001012491.1 | 1 - 2 | 110 | 81 | |
GenBank mRNA | BC089905.1 | - | 1 - 2 | 110 | 81 | |
FQ225242.1 | - | 1 - 2 | 130 | 81 | ||
FQ226282.1 | - | 1 - 2 | 149 | 81 | ||
FQ227427.1 | - | 1 - 2 | 130 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
