Assay Details
Gene Symbol: Mmp15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106168.2 | NP_001099638.1 | 6 - 7 | 1699 | 57 | |
GenBank mRNA | CK357842.1 | - | 1 - 2 | 92 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
