Assay Details
Gene Symbol: Mmp15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106168.2 | NP_001099638.1 | 8 - 9 | 1990 | 125 | |
GenBank mRNA | CK357842.1 | - | 3 - 4 | 383 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
