Assay Details
Gene Symbol: Adk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012895.3 | NP_037027.2 | 1 - 2 | 75 | 72 | |
GenBank mRNA | BC081712.1 | - | 1 - 2 | 75 | 72 | |
U90340.1 | - | 1 - 2 | 78 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
