Assay Details
Gene Symbol: Adk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012895.3 | NP_037027.2 | 3 - 4 | 206 | 74 | |
XM_006251631.1 | XP_006251693.1 | 3 - 4 | 320 | 74 | ||
GenBank mRNA | BC081712.1 | - | 3 - 4 | 206 | 74 | |
U57042.1 | - | 2 - 3 | 146 | 74 | ||
U90340.1 | - | 3 - 4 | 209 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
