Assay Details
Gene Symbol: Adk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012895.3 | NP_037027.2 | 4 - 5 | 282 | 71 | |
XM_006251631.1 | XP_006251693.1 | 4 - 5 | 396 | 71 | ||
GenBank mRNA | BC081712.1 | - | 4 - 5 | 282 | 71 | |
U57042.1 | - | 3 - 4 | 222 | 71 | ||
U90340.1 | - | 4 - 5 | 285 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
