Assay Details
Gene Symbol: Ndst2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105740.1 | NP_001099210.1 | 219 | 154 | ||
XM_008770476.1 | XP_008768698.1 | 1 - 2 | 598 | 154 | ||
XM_008770477.1 | XP_008768699.1 | 1 - 2 | 598 | 154 | ||
XM_008770478.1 | XP_008768700.1 | 1 - 2 | 598 | 154 | ||
XM_008770479.1 | XP_008768701.1 | 1 - 2 | 598 | 154 |
Gene Symbol: NEWGENE_1304700 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006251622.2 | XP_006251684.1 | 1 - 2 | 598 | 154 | |
XM_006251623.2 | XP_006251685.1 | 1 - 2 | 598 | 154 | ||
XM_008770578.1 | XP_008768800.1 | 1 - 2 | 598 | 154 | ||
XM_008770579.1 | XP_008768801.1 | 1 - 2 | 598 | 154 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001105740.1, XM_006251622.2, XM_006251623.2, XM_008770476.1, XM_008770477.1, XM_008770478.1, XM_008770479.1, XM_008770578.1, XM_008770579.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
