Assay Details
Gene Symbol: Ndst2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105740.1 | NP_001099210.1 | 1835 | 68 | ||
XM_008770476.1 | XP_008768698.1 | 3 - 4 | 1999 | 68 | ||
XM_008770477.1 | XP_008768699.1 | 3 - 4 | 1999 | 68 | ||
XM_008770478.1 | XP_008768700.1 | 3 - 4 | 1999 | 68 | ||
XM_008770479.1 | XP_008768701.1 | 3 - 4 | 1999 | 68 |
Gene Symbol: NEWGENE_1304700 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006251622.2 | XP_006251684.1 | 3 - 4 | 1999 | 68 | |
XM_006251623.2 | XP_006251685.1 | 3 - 4 | 1999 | 68 | ||
XM_008770578.1 | XP_008768800.1 | 3 - 4 | 1999 | 68 | ||
XM_008770579.1 | XP_008768801.1 | 3 - 4 | 1999 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
