Assay Details
Gene Symbol: Ndst2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105740.1 | NP_001099210.1 | 3148 | 60 | ||
XM_008770476.1 | XP_008768698.1 | 12 - 13 | 3312 | 60 | ||
XM_008770477.1 | XP_008768699.1 | 12 - 13 | 3312 | 60 | ||
XM_008770480.1 | XP_008768702.1 | 9 - 10 | 1523 | 60 |
Gene Symbol: NEWGENE_1304700 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006251622.2 | XP_006251684.1 | 12 - 13 | 3312 | 60 | |
XM_008770578.1 | XP_008768800.1 | 12 - 13 | 3312 | 60 | ||
XM_008770580.1 | XP_008768802.1 | 9 - 10 | 1523 | 60 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
