Assay Details
Gene Symbol: Tbpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127201.1 | NP_001120673.1 | 3 - 4 | 466 | 78 | |
XM_006227721.2 | XP_006227783.1 | 3 - 4 | 499 | 78 | ||
XM_006227722.2 | XP_006227784.1 | 3 - 4 | 423 | 78 | ||
XM_008758627.1 | XP_008756849.1 | 4 - 5 | 466 | 78 | ||
GenBank mRNA | CF109177.1 | - | 2 - 3 | 158 | 78 | |
FQ213058.1 | - | 2 - 3 | 118 | 78 | ||
FQ220549.1 | - | 2 - 3 | 118 | 78 | ||
FQ234587.1 | - | 3 - 4 | 484 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
