Assay Details
Gene Symbol: Tbpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006227722.2 | XP_006227784.1 | 1 - 2 | 165 | 106 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006227722.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
