Assay Details
Gene Symbol: Papd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107333.1 | NP_001100803.1 | 7 - 8 | 918 | 80 | |
XM_006227781.2 | XP_006227843.1 | 7 - 8 | 4668 | 80 | ||
XM_006227782.2 | XP_006227844.1 | 7 - 8 | 1305 | 80 | ||
XM_008758690.1 | XP_008756912.1 | 7 - 8 | 1035 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
