Assay Details
Gene Symbol: Arg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017134.3 | NP_058830.2 | 5 - 6 | 696 | 107 | |
GenBank mRNA | BC091158.1 | - | 5 - 6 | 626 | 7368991 | 107 |
FQ209619.1 | - | 2 - 3 | 207 | 108 | ||
FQ218591.1 | - | 5 - 6 | 107 | |||
FQ219159.1 | - | 5 - 6 | 577 | 107 | ||
J02720.1 | - | 5 - 6 | 577 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
